Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185064.2 | NP_001171993.1 | 2 - 3 | 194 | 77 | |
XM_013983958.2 | XP_013839412.1 | 866 | 77 | |||
XM_021074510.1 | XP_020930169.1 | 1085 | 77 | |||
XM_021074511.1 | XP_020930170.1 | 832 | 77 | |||
GenBank mRNA | AK237456.1 | - | 3 - 4 | 431 | 77 | |
AK391929.1 | - | 2 - 3 | 513 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |