Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185129.1 | NP_001172058.1 | 2 - 3 | 245 | 57 | |
GenBank mRNA | GU386350.1 | - | 2 - 3 | 245 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |