Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185135.1 | NP_001172064.1 | 3 - 4 | 256 | 107 | |
GenBank mRNA | AK230934.1 | - | 4 - 5 | 310 | 107 | |
EU868836.1 | - | 3 - 4 | 256 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Sus scrofa Probe spans exons |