Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190169.1 | NP_001177098.1 | 6 - 7 | 1200 | 62 | |
GenBank mRNA | AK236562.1 | - | 6 - 7 | 1200 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |