Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTCAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190186.1 | NP_001177115.1 | 4 - 5 | 371 | 60 | |
GenBank mRNA | AK238420.1 | - | 4 - 5 | 315 | 60 | |
AK400712.1 | - | 4 - 5 | 384 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |