Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190203.1 | NP_001177132.1 | 1 - 2 | 51 | 62 | |
XM_005656224.3 | XP_005656281.1 | 144 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |