Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190209.1 | NP_001177138.1 | 1 - 2 | 193 | 94 | |
GenBank mRNA | AK235203.1 | - | 1 - 2 | 174 | 94 | |
BP173023.1 | - | 1 - 2 | 101 | 94 | ||
CV868497.1 | - | 1 - 2 | 193 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |