Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190235.1 | NP_001177164.1 | 4 - 5 | 648 | 89 | |
GenBank mRNA | AK236439.1 | - | 4 - 5 | 651 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |