Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194974.1 | NP_001181903.1 | 11 - 12 | 1403 | 71 | |
XM_021093288.1 | XP_020948947.1 | 1669 | 71 | |||
XM_021093289.1 | XP_020948948.1 | 1764 | 71 | |||
XM_021093290.1 | XP_020948949.1 | 1446 | 71 | |||
XM_021093291.1 | XP_020948950.1 | 1692 | 71 | |||
GenBank mRNA | GQ329716.1 | - | 11 - 12 | 1403 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |