Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195354.1 | NP_001182283.1 | 4 - 5 | 625 | 86 | |
XM_005654294.3 | XP_005654351.1 | 868 | 86 | |||
XM_013983792.2 | XP_013839246.1 | 1479 | 86 | |||
GenBank mRNA | GU373668.1 | - | 4 - 5 | 625 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |