Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195368.1 | NP_001182297.1 | 5 - 6 | 827 | 91 | |
XM_013996752.2 | XP_013852206.1 | 1106 | 91 | |||
GenBank mRNA | AK344212.1 | - | 6 - 7 | 927 | 91 | |
GU373688.1 | - | 5 - 6 | 827 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |