Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199363.1 | NP_001186292.1 | 6 - 7 | 828 | 104 | |
GenBank mRNA | EU617320.1 | - | 6 - 7 | 828 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |