Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204769.1 | NP_001191698.1 | 4 - 5 | 864 | 59 | |
XM_005667404.3 | XP_005667461.1 | 1175 | 59 | |||
XM_005667405.3 | XP_005667462.1 | 1012 | 59 | |||
XM_021102056.1 | XP_020957715.1 | 1155 | 59 | |||
XM_021102057.1 | XP_020957716.1 | 1158 | 59 | |||
XM_021102058.1 | XP_020957717.1 | 1158 | 59 | |||
GenBank mRNA | GU989323.1 | - | 4 - 5 | 864 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |