Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204852.1 | NP_001191781.1 | 2 - 3 | 317 | 91 | |
GenBank mRNA | AK230541.1 | - | 2 - 3 | 274 | 91 | |
FD599314.1 | - | 2 - 3 | 310 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |