Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202461.1 | NP_001189390.1 | 3 - 4 | 302 | 110 | |
XM_021066140.1 | XP_020921799.1 | 1488 | 110 | |||
XM_021066141.1 | XP_020921800.1 | 1489 | 110 | |||
XM_021066142.1 | XP_020921801.1 | 1489 | 110 | |||
GenBank mRNA | HQ834533.1 | - | 3 - 4 | 302 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |