Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244870.1 | NP_001231799.1 | 1 - 2 | 96 | 104 | |
XM_021078518.1 | XP_020934177.1 | 645 | 104 | |||
GenBank mRNA | CF176583.1 | - | 1 - 2 | 96 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |