Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244921.1 | NP_001231850.1 | 8 - 9 | 991 | 106 | |
XM_005667490.3 | XP_005667547.1 | 1022 | 106 | |||
XM_013979554.2 | XP_013835008.1 | 981 | 106 | |||
GenBank mRNA | AK232917.1 | - | 6 - 7 | 613 | 106 | |
AK400963.1 | - | 6 - 7 | 614 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |