Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214093.2 | NP_999258.1 | 1 - 2 | 73 | 60 | |
GenBank mRNA | AK390470.1 | - | 1 - 2 | 72 | 60 | |
FS646243.1 | - | 1 - 2 | 75 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |