Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC4H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243282.1 | NP_001230211.1 | 1 - 2 | 197 | 95 | |
XM_021079379.1 | XP_020935038.1 | 210 | 95 | |||
GenBank mRNA | AK349389.1 | - | 1 - 2 | 145 | 95 | |
FS686862.1 | - | 1 - 2 | 111 | 95 | ||
FS689911.1 | - | 1 - 2 | 197 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |