Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243329.1 | NP_001230258.1 | 4 - 5 | 478 | 115 | |
XM_013992576.2 | XP_013848030.1 | 377 | 115 | |||
XM_021086293.1 | XP_020941952.1 | 1177 | 115 | |||
GenBank mRNA | AK347619.1 | - | 1 - 2 | 76 | 115 | |
BP149699.1 | - | 4 - 5 | 479 | 115 | ||
BP172681.1 | - | 1 - 2 | 74 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |