Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243353.1 | NP_001230282.1 | 4 - 5 | 377 | 61 | |
GenBank mRNA | AK344967.1 | - | 4 - 5 | 377 | 61 | |
AK351608.1 | - | 4 - 5 | 376 | 61 | ||
AK400238.1 | - | 4 - 5 | 363 | 61 | ||
AK400822.1 | - | 4 - 5 | 377 | 61 | ||
FS641076.1 | - | 4 - 5 | 377 | 61 | ||
FS700194.1 | - | 4 - 5 | 374 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |