Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243415.1 | NP_001230344.1 | 6 - 7 | 543 | 94 | |
GenBank mRNA | AK391827.1 | - | 6 - 7 | 557 | 94 | |
AY610180.1 | - | 6 - 7 | 543 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |