Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243436.2 | NP_001230365.1 | 4 - 5 | 629 | 70 | |
GenBank mRNA | CV869126.1 | - | 3 - 4 | 567 | 70 | |
CX056678.1 | - | 1 - 2 | 124 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |