Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243436.2 | NP_001230365.1 | 6 - 7 | 790 | 71 | |
GenBank mRNA | CX056678.1 | - | 3 - 4 | 285 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |