Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243607.2 | NP_001230536.1 | 3 - 4 | 349 | 130 | |
GenBank mRNA | AK344359.1 | - | 3 - 4 | 336 | 130 | |
EW144088.2 | - | 3 - 4 | 390 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |