Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243654.1 | NP_001230583.1 | 2 - 3 | 217 | 88 | |
XM_005671293.3 | XP_005671350.1 | 309 | 88 | |||
GenBank mRNA | AK235945.1 | - | 2 - 3 | 217 | 88 | |
AK347603.1 | - | 2 - 3 | 182 | 88 | ||
AK390487.1 | - | 2 - 3 | 229 | 88 | ||
AY610209.1 | - | 2 - 3 | 170 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |