Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243703.1 | NP_001230632.1 | 5 - 6 | 925 | 56 | |
XM_005673416.3 | XP_005673473.1 | 1077 | 56 | |||
XM_005673418.3 | XP_005673475.1 | 969 | 56 | |||
GenBank mRNA | AK351224.1 | - | 5 - 6 | 925 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |