Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRTO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243715.1 | NP_001230644.1 | 1 - 2 | 72 | 67 | |
XM_021093294.1 | XP_020948953.1 | 50 | 67 | |||
GenBank mRNA | AK240022.1 | - | 1 - 2 | 72 | 67 | |
AK351341.1 | - | 1 - 2 | 72 | 67 | ||
AK398581.1 | - | 1 - 2 | 72 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |