Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSH2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243826.1 | NP_001230755.1 | 5 - 6 | 602 | 72 | |
XM_021077864.1 | XP_020933523.1 | 2595 | 72 | |||
GenBank mRNA | AK240093.1 | - | 5 - 6 | 602 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |