Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244059.1 | NP_001230988.1 | 2 - 3 | 139 | 76 | |
GenBank mRNA | AK343429.1 | - | 2 - 3 | 76 | ||
AK345129.1 | - | 2 - 3 | 112 | 76 | ||
AK396351.1 | - | 2 - 3 | 142 | 76 | ||
BE014473.1 | - | 2 - 3 | 140 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |