Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244070.1 | NP_001230999.1 | 1 - 2 | 293 | 60 | |
XM_005664714.3 | XP_005664771.2 | 182 | 60 | |||
GenBank mRNA | AK233375.1 | - | 1 - 2 | 40 | 60 | |
AK236665.1 | - | 598 | 60 | |||
AK347721.1 | - | 1 - 2 | 41 | 60 | ||
AK392151.1 | - | 1 - 2 | 98 | 60 | ||
AK392193.1 | - | 1 - 2 | 41 | 60 | ||
CJ013638.1 | - | 1 - 2 | 293 | 60 | ||
EW433196.2 | - | 1 - 2 | 82 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |