Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244096.1 | NP_001231025.1 | 4 - 5 | 520 | 100 | |
XM_013986455.2 | XP_013841909.1 | 575 | 100 | |||
XM_021079411.1 | XP_020935070.1 | 592 | 100 | |||
XM_021079412.1 | XP_020935071.1 | 575 | 100 | |||
GenBank mRNA | AK235135.1 | - | 4 - 5 | 520 | 100 | |
AK236383.1 | - | 4 - 5 | 539 | 100 | ||
AK236451.1 | - | 4 - 5 | 523 | 100 | ||
AK236585.1 | - | 4 - 5 | 524 | 100 | ||
AK394814.1 | - | 4 - 5 | 520 | 100 | ||
AK397739.1 | - | 4 - 5 | 575 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |