Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244155.1 | NP_001231084.1 | 5 - 6 | 1029 | 134 | |
XM_005666776.2 | XP_005666833.1 | 1024 | 134 | |||
GenBank mRNA | AK236346.1 | - | 5 - 6 | 1016 | 134 | |
AK395497.1 | - | 5 - 6 | 1029 | 134 | ||
AY609583.1 | - | 5 - 6 | 828 | 134 |
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK391323.1 | - | 2769 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |