Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244163.1 | NP_001231092.1 | 3 - 4 | 454 | 103 | |
XM_021063987.1 | XP_020919646.1 | 488 | 103 | |||
GenBank mRNA | AK234259.1 | - | 3 - 4 | 463 | 103 | |
AK234897.1 | - | 3 - 4 | 432 | 103 | ||
AK351953.1 | - | 3 - 4 | 454 | 103 | ||
AY609413.1 | - | 3 - 4 | 414 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |