Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244185.1 | NP_001231114.1 | 10 - 11 | 1176 | 60 | |
XM_005654367.3 | XP_005654424.1 | 1186 | 60 | |||
XM_021089351.1 | XP_020945010.1 | 1186 | 60 | |||
GenBank mRNA | AK343924.1 | - | 10 - 11 | 1176 | 60 | |
AK346390.1 | - | 11 - 12 | 1136 | 60 | ||
AK351440.1 | - | 10 - 11 | 1066 | 60 | ||
AY609573.1 | - | 10 - 11 | 1101 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |