Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244243.2 | NP_001231172.1 | 1 - 2 | 135 | 55 | |
XM_021068487.1 | XP_020924146.1 | 135 | 55 | |||
GenBank mRNA | AK234347.1 | - | 1 - 2 | 136 | 55 | |
AK391715.1 | - | 1 - 2 | 106 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |