Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244431.1 | NP_001231360.1 | 1 - 2 | 43 | 91 | |
GenBank mRNA | AK238460.1 | - | 1 - 2 | 43 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |