Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244432.1 | NP_001231361.1 | 9 - 10 | 972 | 64 | |
GenBank mRNA | AK232087.1 | - | 9 - 10 | 972 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |