Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244435.1 | NP_001231364.1 | 3 - 4 | 510 | 69 | |
GenBank mRNA | AK234097.1 | - | 3 - 4 | 510 | 69 | |
AY609710.1 | - | 3 - 4 | 448 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |