Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244501.2 | NP_001231430.1 | 10 - 11 | 1526 | 94 | |
GenBank mRNA | AK234901.1 | - | 10 - 11 | 1593 | 94 | |
AK235266.1 | - | 5 - 6 | 522 | 94 | ||
AK235921.1 | - | 10 - 11 | 1201 | 94 | ||
AK343698.1 | - | 7 - 8 | 774 | 94 | ||
AK392294.1 | - | 10 - 11 | 1526 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |