Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244502.1 | NP_001231431.1 | 4 - 5 | 335 | 72 | |
XM_005659420.3 | XP_005659477.2 | 334 | 72 | |||
XM_005659421.3 | XP_005659478.2 | 333 | 72 | |||
XM_005659422.3 | XP_005659479.2 | 331 | 72 | |||
XM_005659423.3 | XP_005659480.2 | 326 | 72 | |||
GenBank mRNA | AK399976.1 | - | 4 - 5 | 276 | 72 | |
FD593970.1 | - | 4 - 5 | 337 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |