Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244548.1 | NP_001231477.1 | 3 - 4 | 356 | 124 | |
GenBank mRNA | AK233495.1 | - | 3 - 4 | 317 | 124 | |
AK391903.1 | - | 3 - 4 | 357 | 124 | ||
AK399059.1 | - | 1 - 2 | 50 | 124 | ||
AY609451.1 | - | 3 - 4 | 316 | 124 | ||
BW976052.1 | - | 3 - 4 | 317 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |