Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244581.1 | NP_001231510.1 | 2 - 3 | 528 | 92 | |
XM_021072058.1 | XP_020927717.1 | 216 | 92 | |||
XM_021072059.1 | XP_020927718.1 | 331 | 92 | |||
GenBank mRNA | FS666888.1 | - | 2 - 3 | 528 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |