Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244639.1 | NP_001231568.1 | 6 - 7 | 833 | 108 | |
GenBank mRNA | AK232700.1 | - | 6 - 7 | 833 | 108 | |
AK232723.1 | - | 6 - 7 | 770 | 108 | ||
AK232888.1 | - | 6 - 7 | 774 | 108 | ||
AK394210.1 | - | 3 - 4 | 292 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |