Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244681.1 | NP_001231610.1 | 4 - 5 | 854 | 97 | |
GenBank mRNA | AK345827.1 | - | 4 - 5 | 854 | 97 | |
AK391863.1 | - | 4 - 5 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |