Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244681.1 | NP_001231610.1 | 6 - 7 | 1112 | 74 | |
GenBank mRNA | AK345827.1 | - | 6 - 7 | 1112 | 74 | |
AK391863.1 | - | 6 - 7 | 1105 | 74 | ||
AK400673.1 | - | 1 - 2 | 74 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |