Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206336.1 | NP_001193265.1 | 2 | 1225 | 145 | |
XM_013983672.2 | XP_013839126.1 | 1664 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |