Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021089893.1 | XP_020945552.1 | 3499 | 102 | ||
GenBank mRNA | AK230540.1 | - | 6 - 7 | 893 | 102 | |
AK239742.1 | - | 6 - 7 | 874 | 102 | ||
AK389756.1 | - | 6 - 7 | 935 | 102 | ||
AK389844.1 | - | 6 - 7 | 935 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |