Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY515506.1 | - | 54 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Triticum aestivum |