Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ237992.1 | - | 5 - 6 | 1401 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Vitis vinifera Probe spans exons |